Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Dimethylbiphenyl 98%

Supplier: AMBEED, INC
Warning

A758981-5G A758981-1G
77255-670EA 45 USD InStock
77255-670 77595-198
2,2'-Dimethylbiphenyl 98%
2,2'-Dimethylbiphenyl
Precaución: for Research Use Only


Formula: C₁₄H₁₄
MW: 182.27 g/mol
Boiling Pt: 258
Melting Pt: 18
MDL Number: MFCD00048075
CAS Number: 605-39-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty