Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

STF-31 98%

Supplier: AMBEED, INC
Warning

A761947-10MG A761947-1MG A761947-50MG A761947-5MG
77243-950EA 31 USD InStock
77243-950 77243-952 77243-954 77243-956
STF-31 98%
STF-31
Precaución: for Research Use Only


Formula: C₂₃H₂₅N₃O₃S
MDL Number: MFCD04153828
CAS Number: 724741-75-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty