Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Butyl-2-ethyl-1,3-propanediol 98%

Supplier: AMBEED, INC
Warning

A763310-100G A763310-25G A763310-500G A763310-1000G
77267-922EA 32 USD InStock
77267-922 77267-924 77267-926 77749-521
2-Butyl-2-ethyl-1,3-propanediol 98%
2-Butyl-2-ethyl-1,3-propanediol
Precaución: for Research Use Only


Formula: C₉H₂₀O₂
MW: 160.26 g/mol
Boiling Pt: 262
Melting Pt: 39
MDL Number: MFCD00004697
CAS Number: 115-84-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty