Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-methoxybenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A765791-100G A765791-1G A765791-25G A765791-5G
77293-602EA 851 USD InStock
77293-602 77293-604 77293-606 77293-608
4-Fluoro-3-methoxybenzyl alcohol 98%
4-Fluoro-3-methoxybenzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₉FO₂
MDL Number: MFCD00272133
CAS Number: 128495-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty