Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluorobenzoyl chloride 98%

Supplier: AMBEED, INC
Danger

A766394-25G A766394-5G
77267-958EA 19 USD InStock
77267-958 77267-960
2,6-Difluorobenzoyl chloride 98%
2,6-Difluorobenzoyl chloride
Precaución: for Research Use Only


Formula: C₇H₃ClF₂O
MDL Number: MFCD00000659
CAS Number: 18063-02-0
UN: 2920
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty