Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-4-methoxybutane 98%

Supplier: AMBEED, INC
Danger

A767437-10G A767437-25G
77243-982EA 5 USD InStock
77243-982 77243-984
1-Chloro-4-methoxybutane 98%
1-Chloro-4-methoxybutane
Precaución: for Research Use Only


Formula: C₅H₁₁ClO
MW: 122.59
MDL Number: MFCD00209961
CAS Number: 17913-18-7
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty