Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,5-diaminopyridine 95%

Supplier: AMBEED, INC
Danger

A768078-100MG A768078-10G A768078-5G
77281-258EA 6 USD InStock
77281-258 77281-260 77281-262
3-Bromo-2,5-diaminopyridine 95%
3-Bromo-2,5-diaminopyridine
Precaución: for Research Use Only


Formula: C₅H₆BrN₃
MDL Number: MFCD03427651
CAS Number: 896160-69-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty