Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(4-biphenylyl)amine 98%

Supplier: AMBEED, INC
Warning

A769987-100MG A769987-1G A769987-250MG
77255-754EA 24 USD InStock
77255-754 77255-756 77255-758
Tris(4-biphenylyl)amine 98%
Tris(4-biphenylyl)amine
Precaución: for Research Use Only


Formula: C₃₆H₂₇N
MDL Number: MFCD30470498
CAS Number: 6543-20-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty