Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylbenzyl phenyl ketone 97%

Supplier: AMBEED, INC
Warning

A771498-1G A771498-25G A771498-5G
77255-768EA 23 USD InStock
77255-768 77255-770 77255-772
4-Methylbenzyl phenyl ketone 97%
4-Methylbenzyl phenyl ketone
Precaución: for Research Use Only


Formula: C₁₅H₁₄O
MDL Number: MFCD00017513
CAS Number: 2430-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty