Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-6-(5-(4-methylpiperazine-1-carbonyl)thiophen-2-yl)-1-naphthaldehyde 97%

Supplier: AMBEED, INC
Warning

A777720-10MG A777720-1MG A777720-25MG A777720-50MG A777720-5MG
77268-060EA 43 USD InStock
77268-060 77268-062 77268-064 77268-066 77268-068
2-Hydroxy-6-(5-(4-methylpiperazine-1-carbonyl)thiophen-2-yl)-1-naphthaldehyde 97%
2-Hydroxy-6-(5-(4-methylpiperazine-1-carbonyl)thiophen-2-yl)-1-naphthaldehyde
Precaución: for Research Use Only


Formula: C₂₁H₂₀N₂O₃S
MDL Number: MFCD30496700
CAS Number: 1093119-54-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty