Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Difluoropyridine-3-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A779366-100MG A779366-1G A779366-5G
77255-792EA 74 USD InStock
77255-792 77255-794 77255-796
2,5-Difluoropyridine-3-carboxylic acid 95%
2,5-Difluoropyridine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₃F₂NO₂
MDL Number: MFCD07777166
CAS Number: 851386-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty