Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Phenylthio)phthalimide 95%

Supplier: AMBEED, INC
Warning

A781522-1G A781522-250MG A781522-5G
77244-092EA 18 USD InStock
77244-092 77244-094 77244-096
N-(Phenylthio)phthalimide 95%
N-(Phenylthio)phthalimide
Precaución: for Research Use Only


Formula: C₁₄H₉NO₂S
MDL Number: MFCD00192396
CAS Number: 14204-27-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty