Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethoxyphenol 96%

Supplier: AMBEED, INC
Warning

A783226-10G A783226-1G A783226-5G
77281-302EA 35 USD InStock
77281-302 77281-304 77281-306
3,4-Dimethoxyphenol 96%
3,4-Dimethoxyphenol
Precaución: for Research Use Only


Formula: C₈H₁₀O₃
MW: 154.17
Melting Pt: 83
MDL Number: MFCD00008390
CAS Number: 2033-89-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty