Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromobenzene-1,4-diamine 97%

Supplier: AMBEED, INC
Warning

A783414-100MG A783414-1G A783414-250MG A783414-5G
77244-110EA 5 USD InStock
77244-110 77244-112 77244-114 77244-116
2,5-Dibromobenzene-1,4-diamine 97%
2,5-Dibromobenzene-1,4-diamine
Precaución: for Research Use Only


Formula: C₆H₆Br₂N₂
MDL Number: MFCD19441210
CAS Number: 25462-61-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty