Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-1H-indole-3-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A783598-1G A783598-250MG A783598-5G
77293-690EA 64 USD InStock
77293-690 77293-692 77293-694
4-Hydroxy-1H-indole-3-carbaldehyde 95%
4-Hydroxy-1H-indole-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MW: 161.1574 g/mol
MDL Number: MFCD05864721
CAS Number: 81779-27-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty