Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenyl-2-butanone 95%

Supplier: AMBEED, INC
Warning

A784621-1G A784621-250MG A784621-5G
77244-118EA 36 USD InStock
77244-118 77244-120 77244-122
1-Phenyl-2-butanone 95%
1-Phenyl-2-butanone
Precaución: for Research Use Only


Formula: C₆H₅C(O)CH₂CH₂CH₃
MW: 148.2 g/mol
Boiling Pt: 230
Melting Pt: 5
MDL Number: MFCD00009315
CAS Number: 1007-32-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty