Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyladamantan-2-yl acrylate 98% + stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A786979-1G A786979-25G A786979-5G
77268-114EA 12 USD InStock
77268-114 77268-116 77268-118
2-Methyladamantan-2-yl acrylate 98% + stabilized
2-Methyladamantan-2-yl acrylate
Precaución: for Research Use Only


Formula: C₁₄H₂₀O₂
MDL Number: MFCD08704223
CAS Number: 249562-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty