Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Isopropylphenyl)isobutyraldehyde 92%

Supplier: AMBEED, INC
Warning

A789477-100G A789477-25G A789477-500G A789477-5G
77281-350EA 81 USD InStock
77281-350 77281-352 77281-354 77281-356
3-(4-Isopropylphenyl)isobutyraldehyde 92%
3-(4-Isopropylphenyl)isobutyraldehyde


Formula: C₁₃H₁₈O
MDL Number: MFCD00024160
CAS Number: 103-95-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty