Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,6-Dichlorophenyl)acetic acid methyl ester 98%

Supplier: AMBEED, INC
Warning

A791197-10G A791197-1G A791197-25G A791197-5G
77268-174EA 51 USD InStock
77268-174 77268-176 77268-178 77268-180
(2,6-Dichlorophenyl)acetic acid methyl ester 98%
(2,6-Dichlorophenyl)acetic acid methyl ester
Precaución: for Research Use Only


Formula: (C₆H₃Cl₂)CH₂COOCH₃
MDL Number: MFCD00191640
CAS Number: 54551-83-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty