Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Amino-2'-hydroxyacetophenone hydrochloride 98%

Supplier: AMBEED, INC
Warning

A791694-100G A791694-25G A791694-5G
77255-886EA 144 USD InStock
77255-886 77255-888 77255-890
3'-Amino-2'-hydroxyacetophenone hydrochloride 98%
3'-Amino-2'-hydroxyacetophenone hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₀ClNO₂
MDL Number: MFCD03790987
CAS Number: 90005-55-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty