Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bentamapimod 98%

Supplier: AMBEED, INC
Warning

A793130-10MG A793130-1MG A793130-25MG A793130-5MG A793130-50MG
77244-160EA 63 USD InStock
77244-160 77244-162 77244-164 77244-166 77747-545
Bentamapimod 98%
Bentamapimod
Precaución: for Research Use Only


Formula: C₂₅H₂₃N₅O₂S
MDL Number: MFCD08272350
CAS Number: 848344-36-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty