Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,1,3-Benzoxadiazole-4-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A794124-1G A794124-250MG A794124-25G A794124-5G
77281-378EA 37 USD InStock
77281-378 77281-380 77281-382 77281-384
2,1,3-Benzoxadiazole-4-carbaldehyde 98%
2,1,3-Benzoxadiazole-4-carbaldehyde
Precaución: for Research Use Only


Formula: C₇H₄N₂O₂
MDL Number: MFCD07772900
CAS Number: 32863-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty