Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(difluoromethyl)nicotinate 97%

Supplier: AMBEED, INC
Warning

A794852-100MG A794852-1G A794852-250MG A794852-50MG
77268-200EA 42 USD InStock
77268-200 77268-202 77268-204 77268-206
Ethyl 2-(difluoromethyl)nicotinate 97%
Ethyl 2-(difluoromethyl)nicotinate
Precaución: for Research Use Only


Formula: C₉H₉F₂NO₂
MDL Number: MFCD25476644
CAS Number: 1346541-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty