Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3,3'-Bis(9-anthracenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl HydrogenPhosphate 98%

Supplier: AMBEED, INC
Warning

A795910-100MG A795910-1G A795910-250MG
77255-908EA 55 USD InStock
77255-908 77255-910 77255-912
(R)-3,3'-Bis(9-anthracenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl HydrogenPhosphate 98%
(R)-3,3'-Bis(9-anthracenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl HydrogenPhosphate
Precaución: for Research Use Only


Formula: C₄₈H₃₇O₄P
MDL Number: MFCD30489118
CAS Number: 1011465-29-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty