Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine 97%

Supplier: AMBEED, INC
Warning

A800994-1G A800994-250MG A800994-5G
77293-796EA 31 USD InStock
77293-796 77293-798 77293-800
1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine 97%
1-tert-Butyloxycarbonyl-4-(3-aminopropyl)piperazine
Precaución: for Research Use Only


Formula: C₁₂H₂₅N₃O₂
MDL Number: MFCD06808578
CAS Number: 373608-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty