Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

IMR-1 97%

Supplier: AMBEED, INC
Warning

A802072-10MG A802072-1MG A802072-50MG A802072-5MG
77268-250EA 32 USD InStock
77268-250 77268-252 77268-254 77268-256
IMR-1 97%
IMR-1
Precaución: for Research Use Only


Formula: C₁₅H₁₅NOS₂
MDL Number: MFCD04141288
CAS Number: 310456-65-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty