Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzoyl-5-bromothiophene 97%

Supplier: AMBEED, INC
Warning

A803232-100G A803232-10G A803232-1G A803232-250MG A803232-25G A803232-5G
77268-262EA 450.01 USD InStock
77268-262 77268-264 77268-266 77268-268 77268-270 77268-272
2-Benzoyl-5-bromothiophene 97%
2-Benzoyl-5-bromothiophene
Precaución: for Research Use Only


Formula: C₁₁H₇BrOS
MDL Number: MFCD03086026
CAS Number: 31161-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty