Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2'-(dicyclohexylphosphino)-2,6-dimethoxy-[1,1'-biphenyl]-3-sulfonate 97%

Supplier: AMBEED, INC
Warning

A803754-5G A803754-1G
77268-274EA 114 USD InStock
77268-274 77749-590
Sodium 2'-(dicyclohexylphosphino)-2,6-dimethoxy-[1,1'-biphenyl]-3-sulfonate 97%
Sodium 2'-(dicyclohexylphosphino)-2,6-dimethoxy-[1,1'-biphenyl]-3-sulfonate
Precaución: for Research Use Only


Formula: C₂₆H₃₄NaO₅PS
MDL Number: MFCD14702646
CAS Number: 870245-75-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty