Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroethyl phenyl sulfide 98%

Supplier: AMBEED, INC
Danger

A804851-100G A804851-10G A804851-1G A804851-25G A804851-5G
77268-292EA 180 USD InStock
77268-292 77268-294 77268-296 77268-298 77268-300
2-Chloroethyl phenyl sulfide 98%
2-Chloroethyl phenyl sulphide
Precaución: for Research Use Only


Formula: C₈H₉ClS
MW: 172.68 g/mol
MDL Number: MFCD00013691
CAS Number: 5535-49-9
UN: 2810
ADR: 6.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty