Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-(trifluoromethyl)anisole 98%

Supplier: AMBEED, INC
Warning

A805613-100G A805613-1G A805613-25G A805613-500G A805613-5G
77293-802EA 164 USD InStock
77293-802 77293-804 77293-806 77293-808 77293-810
4-Bromo-2-(trifluoromethyl)anisole 98%
4-Bromo-2-(trifluoromethyl)anisole
Precaución: for Research Use Only


Formula: C₈H₆BrF₃O
MDL Number: MFCD01631579
CAS Number: 1514-11-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty