Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4,5-difluorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A809332-1G A809332-250MG A809332-5G
77268-310EA 36 USD InStock
77268-310 77268-312 77268-314
3-Chloro-4,5-difluorobenzonitrile 98%
3-Chloro-4,5-difluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₂ClF₂N
MDL Number: MFCD18415562
CAS Number: 103879-29-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty