Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Naphthaldehyde 98%

Supplier: AMBEED, INC
Warning

A810340-100G A810340-25G A810340-500G A810340-10G
77281-422EA 83 USD InStock
77281-422 77281-424 77281-426 77552-496
2-Naphthaldehyde 98%
2-Naphthaldehyde
Precaución: For research use only.


Formula: C₁₁H₈O
MDL Number: MFCD00004094
CAS Number: 66-99-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty