Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,3,4,4-tetrafluoro-1-butanol 98%

Supplier: AMBEED, INC
Warning

A812526-1G A812526-25G A812526-5G
77293-856EA 22 USD InStock
77293-856 77293-858 77293-860
4-Bromo-3,3,4,4-tetrafluoro-1-butanol 98%
4-Bromo-3,3,4,4-tetrafluoro-1-butanol
Precaución: for Research Use Only


Formula: C₄H₅BrF₄O
MDL Number: MFCD00077467
CAS Number: 234443-21-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty