Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PX-478 hydrochloride 98%

Supplier: AMBEED, INC
Warning

A812611-10MG A812611-50MG
77244-276EA 248 USD InStock
77244-276 77244-278
PX-478 hydrochloride 98%
PX-478 hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₂₀Cl₄N₂O
MDL Number: MFCD11840749
CAS Number: 685898-44-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty