Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-3-butenenitrile 75% (by GC)

Supplier: AMBEED, INC
Danger

A816168-25G A816168-5G
77281-446EA 30 USD InStock
77281-446 77281-448
2-Methyl-3-butenenitrile 75% (by GC)
2-Methyl-3-butenenitrile
Precaución: for Research Use Only


Formula: C₅H₇N
MW: 81.12 g/mol
MDL Number: MFCD00042634
CAS Number: 16529-56-9
UN: 3273
ADR: 3,6.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty