Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Diethylamino-furan-2-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A818521-100MG A818521-1G A818521-250MG
77256-096EA 25 USD InStock
77256-096 77256-098 77256-100
5-Diethylamino-furan-2-carbaldehyde 95%
5-Diethylamino-furan-2-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₃NO₂
MW: 167.205 g/mol
MDL Number: MFCD02614961
CAS Number: 22868-59-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty