Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide 98%

Supplier: AMBEED, INC
Danger

A818722-25G A818722-5G
77244-312EA 19 USD InStock
77244-312 77244-314
1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide 98%
1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide
Precaución: for Research Use Only


Formula: C₈H₁₁F₆N₃O₄S₂
MDL Number: MFCD03788927
CAS Number: 174899-82-2
UN: 2922
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty