Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitrobenzophenone 98%

Supplier: AMBEED, INC
Warning

A821062-1G A821062-250MG A821062-25G A821062-5G
77293-880EA 32 USD InStock
77293-880 77293-882 77293-884 77293-886
3-Nitrobenzophenone 98%
3-Nitrobenzophenone
Precaución: for Research Use Only


Formula: C₁₃H₉NO₃
MDL Number: MFCD00007257
CAS Number: 2243-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty