Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Octen-3-yl Acetate 95%

Supplier: AMBEED, INC
Warning

A823944-100G A823944-25G A823944-5G
77244-340EA 74 USD InStock
77244-340 77244-342 77244-344
1-Octen-3-yl Acetate 95%
1-Octen-3-yl Acetate
Precaución: for Research Use Only


Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
MDL Number: MFCD00036568
CAS Number: 2442-10-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty