Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Perfluoro-n-hexyl)-1,2-propenoxide 98%

Supplier: AMBEED, INC
Danger

A826084-1G A826084-25G A826084-5G
77293-904EA 18 USD InStock
77293-904 77293-906 77293-908
3-(Perfluoro-n-hexyl)-1,2-propenoxide 98%
3-(Perfluoro-n-hexyl)-1,2-propenoxide
Precaución: for Research Use Only


Formula: C₉H₅F₁₃O
MW: 376.12 g/mol
MDL Number: MFCD00042356
CAS Number: 38565-52-5
UN: 1993
ADR: 3 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty