Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Tetrahydrofuroyl)piperazine hydrobromide 97%

Supplier: AMBEED, INC
Warning

A827589-1G A827589-25G A827589-5G
77256-164EA 18 USD InStock
77256-164 77256-166 77256-168
1-(2-Tetrahydrofuroyl)piperazine hydrobromide 97%
1-(2-Tetrahydrofuroyl)piperazine hydrobromide
Precaución: for Research Use Only


Formula: C₉H₁₇BrN₂O₂
MDL Number: MFCD03701619
CAS Number: 63590-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty