Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MK-1064 97%

Supplier: AMBEED, INC
Warning

A827816-10MG A827816-25MG A827816-50MG A827816-5MG
77268-432EA 70 USD InStock
77268-432 77268-434 77268-436 77268-438
MK-1064 97%
MK-1064
Precaución: for Research Use Only


Formula: C₂₄H₂₀ClN₅O₃
MDL Number: MFCD28502027
CAS Number: 1207253-08-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty