Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chlorofuro[3,2-d]pyrimidine 97%

Supplier: AMBEED, INC
Warning

A828888-100MG A828888-25MG A828888-50MG
77268-440EA 410.01 USD InStock
77268-440 77268-442 77268-444
2-Chlorofuro[3,2-d]pyrimidine 97%
2-Chlorofuro[3,2-d]pyrimidine
Precaución: for Research Use Only


Formula: C₆H₃ClN₂O
MDL Number: MFCD22398118
CAS Number: 1643142-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty