Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiazole-4,5-dicarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A831488-100MG A831488-1G A831488-250MG
77244-392EA 111 USD InStock
77244-392 77244-394 77244-396
Thiazole-4,5-dicarboxylic acid 95%
Thiazole-4,5-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₅H₃NO₄S
MW: 173.14 g/mol
MDL Number: MFCD18088933
CAS Number: 22358-80-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty