Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Ethyl-3-(5-phenyl-1H-1,2,3-triazol-1-yl)-9H-carbazole 98%

Supplier: AMBEED, INC
Warning

A832050-10MG A832050-1MG A832050-25MG A832050-5MG
77268-466EA 88 USD InStock
77268-466 77268-468 77268-470 77268-472
9-Ethyl-3-(5-phenyl-1H-1,2,3-triazol-1-yl)-9H-carbazole 98%
9-Ethyl-3-(5-phenyl-1H-1,2,3-triazol-1-yl)-9H-carbazole
Precaución: for Research Use Only


Formula: C₂₂H₁₈N₄
MDL Number: MFCD31746894
CAS Number: 2097938-73-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty