Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Ethylcarbamoyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A837165-1G A837165-250MG A837165-5G
77281-630EA 113 USD InStock
77281-630 77281-632 77281-634
3-(Ethylcarbamoyl)phenylboronic acid 98%
3-(Ethylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
MDL Number: MFCD04115689
CAS Number: 850567-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty