Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Jatrorrhizine 98%

Supplier: AMBEED, INC
Danger

A840009-10MG A840009-1MG A840009-25MG A840009-2MG A840009-5MG A840009-250MG
77256-228EA 35 USD InStock
77256-228 77256-230 77256-232 77256-234 77256-236 77747-601
Jatrorrhizine 98%
Jatrorrhizine
Precaución: for Research Use Only


Formula: C₂₀H₂₀NO₄
MDL Number: MFCD01310242
CAS Number: 3621-38-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty