Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethyl-1H-indole-5-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A840095-100MG A840095-1G A840095-250MG
77256-238EA 53 USD InStock
77256-238 77256-240 77256-242
2,3-Dimethyl-1H-indole-5-carboxylic acid 95%
2,3-Dimethyl-1H-indole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₂
MDL Number: MFCD00464053
CAS Number: 14844-73-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty