Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-pyrrolidin-1-ylpyridine 98%

Supplier: AMBEED, INC
Warning

A843482-1G A843482-250MG A843482-5G
77268-548EA 41 USD InStock
77268-548 77268-550 77268-552
2-Bromo-6-pyrrolidin-1-ylpyridine 98%
2-Bromo-6-pyrrolidin-1-ylpyridine
Precaución: for Research Use Only


Formula: C₉H₁₁BrN₂
MDL Number: MFCD08271892
CAS Number: 230618-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty