Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-3-hydroxy-6-methylpyridine 95%

Supplier: AMBEED, INC
Warning

A843927-100MG A843927-1G A843927-250MG A843927-5G
77256-254EA 26 USD InStock
77256-254 77256-256 77256-258 77256-260
2,4-Dibromo-3-hydroxy-6-methylpyridine 95%
2,4-Dibromo-3-hydroxy-6-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅Br₂NO
MDL Number: MFCD09753265
CAS Number: 23003-29-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty